Skip to Content
Merck
All Photos(2)

Key Documents

8.45103

Sigma-Aldrich

2-Methyltetrahydrofuran

(stabilised with 2,6-di-tert-butyl-4-methylphenol) Msynthplus

Synonym(s):

Tetrahydro-2-methylfuran, 2-MeTHF, Tetrahydrosilvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102448
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-507-4
bp:
78-80 °C (lit.)
vapor pressure:
136 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

136 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

270 °C

potency

300-2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

1.2-5.7 % (v/v)

dilution

(for synthesis)

refractive index

n20/D 1.406 (lit.)

bp

78-80 °C (lit.)

mp

-136 °C

transition temp

flash point -11 °C

density

0.86 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC1CCCO1

InChI

1S/C5H10O/c1-5-3-2-4-6-5/h5H,2-4H2,1H3

InChI key

JWUJQDFVADABEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Color (visual): colourless
Appearance of substance (visual): clear
Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.852 - 0.855
Water (K. F.): ≤ 0.01 %
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichlorethane: not measurable
Benzene: not measurable
Carbon tetrachloride: not measurable
1,2-Dichlorethane: not measurable
The material is stabilised with Butylhydroxytoluene (BHT).
At time of analytical testing a content of >100 ppm stabiliser was determined.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(C)

-10.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service