Skip to Content
Merck
All Photos(1)

Documents

8.45087

Sigma-Aldrich

Acetaldehyde dimethyl acetal

Msynthplus

Synonym(s):

Acetaldehyde dimethyl acetal, Dimethyl acetal, 1,1-Dimethoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OCH3)2
CAS Number:
Molecular Weight:
90.12
MDL number:
UNSPSC Code:
12352005
EC Index Number:
208-589-8

vapor pressure

200 hPa ( 20 °C)

Assay

≥96.0% (GC)

form

liquid

potency

6500 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

bp

63-65 °C/1013 hPa

mp

-113 °C

transition temp

flash point -17 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H10O2/c1-4(5-2)6-3/h4H,1-3H3

InChI key

SPEUIVXLLWOEMJ-UHFFFAOYSA-N

Analysis Note

Colour (visual): colourless
Appearance of substance (visual): clear
Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.850 - 0.853
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Methanol (GC): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service