Skip to Content
Merck

Skip To

8.22011

Dehydroacetic acid

for synthesis

Synonym(s):

Dehydroacetic acid, 3-Acetyl-3,4-dihydro-6-methyl-2H-pyran-2,4-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
UNSPSC Code:
12352115
EC Index Number:
208-293-9
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (acidimetric)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.001 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (acidimetric)

form

powder

autoignition temp.

336 °C

potency

1000 mg/kg LD50, oral (Rat)

pH

4 (20 °C, 2 g/L in H2O)

bp

270 °C/1013 hPa

mp

109-111 °C

transition temp

flash point 157 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

O1C(=CC(=O)C(C1=O)C(=O)C)C

InChI

1S/C8H8O4/c1-4-3-6(10)7(5(2)9)8(11)12-4/h3,7H,1-2H3

InChI key

PGRHXDWITVMQBC-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 109 °C
Melting range (upper value): ≤ 112 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.140858.047848.02971
assay

≥99.0% (acidimetric)

assay

-

assay

≥97.0% (acidimetric)

assay

≥98.0% (GC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

bp

270 °C/1013 hPa

bp

-

bp

350 °C/1013 hPa

bp

85.5 °C/1013 hPa

form

powder

form

powder

form

powder

form

liquid

mp

109-111 °C

mp

280-285 °C

mp

230-232 °C

mp

-70 °C


Still not finding the right product?

Explore all of our products under Dehydroacetic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

314.6 °F

flash_point_c

157 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters