Skip to Content
Merck
All Photos(1)

Key Documents

8.21130

Sigma-Aldrich

4-Methylthiophenol

for synthesis

Synonym(s):

4-Methylthiophenol, p-Tolyl mercaptan, p-Thiocresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3)C6H4SH
CAS Number:
Molecular Weight:
124.20
MDL number:
UNSPSC Code:
12352105
EC Index Number:
203-399-1
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

powder

bp

195 °C/1013 hPa

mp

39-42 °C

transition temp

flash point 70 °C

storage temp.

2-30°C

SMILES string

Sc1ccc(cc1)C

InChI

1S/C7H8S/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3

InChI key

WLHCBQAPPJAULW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 39 °C
Melting range (upper value): ≤ 42 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service