Skip to Content
Merck
All Photos(1)

Documents

8210-OP

Millipore

Sodium Phosphate

OmniPur® Grade, >= 98.0%

Synonym(s):

Sodium phosphate dibasic heptahydrate, Disodium hydrogen phosphate heptahydrate, Disodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

vapor density

4.9 (vs air)

Quality Level

form

crystalline powder (Fine)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

density

1.68 g/mL at 25 °C (lit.)

shipped in

ambient

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.7H2O/c;;1-5(2,3)4;;;;;;;/h;;(H3,1,2,3,4);7*1H2/q2*+1;;;;;;;;/p-2

InChI key

PYLIXCKOHOHGKQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description



Molecular biology grade.

Specifications:
Arsenic (As): 0.0005% max
Assay: 98.0% min
Chloride (Cl): 0.001% max
DNase: None detected
Heavy metals (as Pb): 0.001% max
Insoluble matter: 0.005% max
Iron (Fe): 0.001% max
Moisture (H2O): 43-50%
Nitrogen compounds (as N): 0.001% max

pH (5%, water): 8.7-9.3
Protease: None detected
RNase: None detected
Sulfate (SO4): 0.005% max





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service