Skip to Content
Merck
All Photos(1)

Key Documents

8.21077

Sigma-Aldrich

o-Terphenyl

for synthesis

Synonym(s):

o-Terphenyl, 1,2-Diphenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C6H5)C6H4C6H5
CAS Number:
Molecular Weight:
230.30
MDL number:
UNSPSC Code:
12352005
EC Index Number:
201-517-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 Pa ( 25 °C)

Quality Level

Assay

≥99.0% (HPLC)

potency

1900 mg/kg LD50, oral (Rat)

bp

332 °C/1013 hPa

mp

55-59 °C

transition temp

flash point 163 °C

storage temp.

2-30°C

SMILES string

c2(c(cccc2)c3ccccc3)c1ccccc1

InChI

1S/C18H14/c1-3-9-15(10-4-1)17-13-7-8-14-18(17)16-11-5-2-6-12-16/h1-14H

InChI key

OIAQMFOKAXHPNH-UHFFFAOYSA-N

Application

  • Hybrid Edge Results in Narrowed Band Gap: Bottom-up Liquid-Phase Synthesis of Bent N = 6/8 Armchair Graphene Nanoribbons.: This study demonstrates a bottom-up liquid-phase synthesis of bent armchair graphene nanoribbons with a hybrid edge structure, resulting in a narrowed band gap. The application of o-Terphenyl in this synthesis is crucial for achieving the desired properties of the nanoribbons (Li et al., 2024).
  • White Light from Blue Fluorescence and Sensitized Yellow Long-Afterglow Phosphorescence of o-Terphenyl in Its ο-Acid···Base Adduct with Ag(3)Pz(3).: The research explores the optical properties of o-Terphenyl in a complex with Ag(3)Pz(3), showing its potential in white light generation through blue fluorescence and yellow long-afterglow phosphorescence (Zhan et al., 2019).
  • Regularized Dynamical Decoupling Noise Spectroscopy - A Decoherence Descriptor for Radicals in Glassy Matrices.: This paper describes the use of o-Terphenyl as a glassy matrix for noise spectroscopy, providing insights into the decoherence processes of radicals. The findings highlight o-Terphenyl′s role in stabilizing radicals for advanced spectroscopic applications (Soetbeer et al., 2021).
  • Counterion Influence on Dynamic Spin Properties in a V(iv) Complex.: The influence of counterions on the dynamic spin properties of a V(iv) complex is investigated, with o-Terphenyl playing a role in modulating the magnetic properties of the complex. This study underscores the material′s significance in magnetic resonance applications (Lin et al., 2019).

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

325.4 °F

Flash Point(C)

163 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service