Skip to Content
Merck
All Photos(1)

Documents

8.20661

Sigma-Aldrich

4-Hydroxybenzophenone

for synthesis

Synonym(s):

4-Hydroxybenzophenone, p-Benzoylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(OH)C6H4COC6H5
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352115
EC Index Number:
214-507-1
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

260-262 °C/32 hPa

mp

131-133 °C

storage temp.

2-30°C

InChI

1S/C13H10O2/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,14H

InChI key

NPFYZDNDJHZQKY-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 131 °C
Melting range (upper value): ≤ 133 °C
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service