Skip to Content
Merck
All Photos(1)

Documents

8.20553

Sigma-Aldrich

2,3-Epoxypropyl phenyl ether

for synthesis

Synonym(s):

2,3-Epoxypropyl phenyl ether, Glycidyl phenyl ether, 1,2-Epoxy-3-phenoxypropane, Phenyl 2,3-epoxypropyl ether, Phenyl glycid ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352112
EC Index Number:
204-557-2
NACRES:
NA.22

vapor pressure

0.013 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

430 °C

potency

2150 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)

mp

3.5 °C

transition temp

flash point 114 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H10O2/c1-2-4-8(5-3-1)10-6-9-7-11-9/h1-5,9H,6-7H2

InChI key

FQYUMYWMJTYZTK-UHFFFAOYSA-N

Application

  • Synthesis of chiral cyclic carbonates via kinetic resolution of racemic epoxides: Focuses on kinetic resolution processes involving 2,3-epoxypropyl phenyl ether, contributing to advancements in chiral compound synthesis (Wu et al., 2016) (MDPI Link).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.107 - 1.111
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service