820453
Probumin® Bovine Serum Albumin Diagnostic Grade
USA origin, Tested for IgG, enzyme immunoassay: suitable, ELISA: suitable, Serology: suitable
About This Item
Recommended Products
product name
Probumin® Bovine Serum Albumin Diagnostic Grade, lyophilized powder, pkg of 10 g
grade
special quality (reovirus, rabies, bov adenovirus, bov parvovirus, bov RSV: none detected)
Quality Level
Assay
≥98% (purity)
form
lyophilized powder
shelf life
48 mo.
mol wt
~66 kDa
purified by
heat shock fractionation
packaging
pkg of 10 g
manufacturer/tradename
Probumin®
origin
USA origin
technique(s)
ELISA: suitable
enzyme immunoassay: suitable
dilution
(Serology)
impurities
Tested for IgG
loss
≤5%
pH
6.8-7.2
anion traces
chloride (Cl-): 0-3.0 mg/g
cation traces
Na: 0-15.0 mg/g
foreign activity
Blue Tongue, none detected
Tested for Protease
Tested for protease
Vesicular Stomatitis Virus, none detected
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Related Content
Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service