Skip to Content
Merck
All Photos(1)

Documents

8.20261

Sigma-Aldrich

8-Hydroxyquinoline

for synthesis

Synonym(s):

8-Hydroxyquinoline, Oxine, 8-Quinolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-711-1
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

solid

potency

1200 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Addition Reactions

bp

267 °C/1013 hPa

mp

72.5-74.0 °C

solubility

555 mg/L

density

1.034 g/cm3 at 20 °C

application(s)

peptide synthesis

storage temp.

2-30°C

InChI

1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H

InChI key

MCJGNVYPOGVAJF-UHFFFAOYSA-N

Application

<ul>
<li><strong>8-Hydroxyquinoline in luminescent cocrystals for analytical detection:</strong> 8-Hydroxyquinoline is used as an electron donor during the development of luminescent organic cocrystal for the detection of 2,4-dinitroaniline, demonstrating its utility in analytical chemistry applications (Liu et al., 2024).</li>
<li><strong>Extraction and determination of pesticides using 8-Hydroxyquinoline functionalized nanocomposites:</strong> Poly(8-hydroxyquinoline) were utilized for the extraction and determination of organophosphorus pesticides in various environmental samples, showcasing its application in environmental monitoring (Shahsavani and Fakhari, 2024).</li>
<li><strong>8-Hydroxyquinoline in medical imaging and therapy:</strong> 8-Hydroxyquinoline is used as a Schiff base derivative and an auxiliary ligand in the synthesis of the dinuclear lanthanide complex [Ho2(L)2(acac)2(CH3OH)2] (Cong et al., 2024).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 71 °C
Melting range (upper value): ≤ 74 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service