Skip to Content
Merck
All Photos(1)

Key Documents

8.20002

Sigma-Aldrich

Acetaldehyde dimethyl acetal

for synthesis

Synonym(s):

Acetaldehyde dimethyl acetal, Dimethyl acetal, 1,1-Dimethoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OCH3)2
CAS Number:
Molecular Weight:
90.12
MDL number:
UNSPSC Code:
12352112
EC Index Number:
208-589-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

200 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

liquid

potency

6500 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

bp

63-65 °C/1013 hPa

mp

-113 °C

transition temp

flash point -17 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(OC)C)C

InChI

1S/C4H10O2/c1-4(5-2)6-3/h4H,1-3H3

InChI key

SPEUIVXLLWOEMJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.850 - 0.853
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service