Skip to Content
Merck
All Photos(1)

Documents

8.18818

Sigma-Aldrich

3-(Trimethoxysilyl)-1-propanethiol

for synthesis

Synonym(s):

3-(Trimethoxysilyl)-1-propanethiol, (3-Mercaptopropyl)trimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16O3SSi
CAS Number:
Molecular Weight:
196.34
MDL number:
UNSPSC Code:
12352103
EC Index Number:
224-588-5
NACRES:
NA.22

vapor pressure

0.2 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

235 °C

potency

774 mg/kg LD50, oral (Rat)
2268 mg/kg LD50, skin (Rabbit)

bp

213-215 °C/1013 hPa

mp

<-50 °C

transition temp

flash point 85 °C

density

1.05 g/cm3 at 25 °C

storage temp.

2-30°C

InChI

1S/C6H16O3SSi/c1-7-11(8-2,9-3)6-4-5-10/h10H,4-6H2,1-3H3

InChI key

UUEWCQRISZBELL-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.055 - 1.057
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service