Skip to Content
Merck
All Photos(2)

Key Documents

8.18665

Sigma-Aldrich

(R)-(+)-Pulegone

for synthesis

Synonym(s):

(R)-(+)-Pulegone, (R)-(+)-p-Menth-4(8)-en-3-one, (R)-(+)-2-Isopropylidene-5-methylcyclohexanone, (R)-(+)-5-Methyl-2-isopropylidenecyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-943-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.16 hPa ( 25 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

potency

470 mg/kg LD50, oral (Rat)
3090 mg/kg LD50, skin (Rabbit)

mp

≤25 °C

transition temp

flash point 82 °C

density

0.935 g/m3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C1C[C@@H](CCC1=C(C)C)C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (20/4 °C): 0.936 - 0.942
Spec. rotation [α²0/D (undiluted): +21.0 - +24.0 °
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service