Skip to Content
Merck
All Photos(1)

Key Documents

8.18568

Sigma-Aldrich

Undecanoic acid

for synthesis

Synonym(s):

Undecanoic acid, 1-Decanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9COOH
CAS Number:
Molecular Weight:
186.29
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-964-2
Pricing and availability is not currently available.

Quality Level

form

solid

expl. lim.

0.6 % (v/v)

bp

284 °C/1013 hPa

mp

26-29 °C

transition temp

flash point >112 °C

solubility

insoluble 0.05 g/L

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)

InChI key

ZDPHROOEEOARMN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

> 112 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service