8.18028
Formic acid-triethylamine 5:2 addition compound
for synthesis
Synonym(s):
Formic acid-triethylamine 5:2 addition compound
About This Item
Recommended Products
Quality Level
form
liquid
reaction suitability
reagent type: reductant
transition temp
flash point 110 °C
density
1.03 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/2C6H15N.5CH2O2/c2*1-4-7(5-2)6-3;5*2-1-3/h2*4-6H2,1-3H3;5*1H,(H,2,3)
InChI key
NLOIHFJXBNFDSF-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.034 - 1.036
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service