Skip to Content
Merck

Skip To

8.16005

(S)-(+)-Histidine hydrochloride monohydrate

for synthesis

Synonym(s):

L-Histidine monohydrochloride monohydrate, His, L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
UNSPSC Code:
12352107
NACRES:
NA.22
EC Index Number:
211-438-9
MDL number:
Beilstein/REAXYS Number:
4168261
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

crystals

pH

3.5-4.5 (20 °C, 100 g/L in H2O)

mp

254 °C (dec.) (lit.)

solubility

149.55 g/L

density

1.49 g/cm3 at 20 °C

bulk density

770 kg/m3

storage temp.

2-30°C

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Spec. rotation [α²0/D (c=11 in HCl 6 mol/l): +8.5 - +9.5 °
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H07550001.04354H4036
form

crystals

form

-

form

solid

form

powder

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-

Quality Level

200

Quality Level

-

Quality Level

500

Quality Level

400

mp

254 °C (dec.) (lit.)

mp

254 °C (dec.) (lit.)

mp

254 °C (dec.) (lit.)

mp

254 °C (dec.) (lit.)

solubility

149.55 g/L

solubility

-

solubility

149.55 g/L

solubility

H2O: 50 mg/mL

bulk density

770 kg/m3

bulk density

-

bulk density

-

bulk density

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
816005002504022536423371
816005010004022536423388

Questions

Reviews

No rating value

Active Filters