Skip to Content
Merck
All Photos(1)

Key Documents

8.14784

Sigma-Aldrich

Borontrifluoride-acetonitrile complex

for synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
Pricing and availability is not currently available.

vapor pressure

85 Pa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

524 °C (Information refers to the main component.)

expl. lim.

3-17 % (v/v) Information refers to the main component.)

bp

82 °C/1013 hPa

transition temp

flash point 10 °C

density

0.875 g/cm3 at 20 °C

Analysis Note

Assay BF3 (acidimetric): 15.0 - 19.0 %
Density (d 20 °C/ 4 °C): 0.870 - 0.880

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service