Skip to Content
Merck
All Photos(1)

Key Documents

8.14186

Sigma-Aldrich

Glycolic acid

(70% solution in water) for synthesis

Synonym(s):

Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.22
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

form

liquid

pH

0.1 (25 °C in H2O)

bp

112 °C/1013 hPa

mp

10 °C

density

1.262 g/cm3 at 20 °C

storage temp.

2-30°C

Application

  • Poly (lactic-co-glycolic acid)-based composite bone-substitute materials: Focuses on the use of glycolic acid in composite materials for bone substitution and its impact on mechanical properties and degradation performance (D Zhao, T Zhu, J Li, L Cui, Z Zhang, X Zhuang, J Ding - Bioactive Materials, 2021). Read more.
  • An Overview of Poly(lactic-co-glycolic) Acid (PLGA)-Based Biomaterials for Bone Tissue Engineering: Discusses the synthesis and physicochemical characteristics of PLGA composites containing glycolic acid (P Gentile, V Chiono, I Carmagnola - International Journal of Molecular Sciences, 2014). Read more.

Analysis Note

Assay (acidimetric): 67.0 - 73.0 %
Density (d 20 °C/ 4 °C): 1.257 - 1.264
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service