Skip to Content
Merck
All Photos(2)

Key Documents

8.14029

Sigma-Aldrich

(4-Pyridylthio)acetic acid

for synthesis

Synonym(s):

(4-Pyridylthio)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2S
CAS Number:
Molecular Weight:
169.20
MDL number:
UNSPSC Code:
12352106
EC Index Number:
233-765-6
NACRES:
NA.22

Quality Level

Assay

≥98.0% (acidimetric)

autoignition temp.

305 °C

mp

255-258 °C (decomposes)

bulk density

340‑390 kg/m3

storage temp.

2-30°C

SMILES string

S(CC(=O)[O-])c1ccncc1

InChI

1S/C7H7NO2S/c9-7(10)5-11-6-1-3-8-4-2-6/h1-4H,5H2,(H,9,10)/p-1

InChI key

PGUPJAPHYIEKLT-UHFFFAOYSA-M

Related Categories

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service