Skip to Content
Merck
All Photos(1)

Key Documents

8.12034

Sigma-Aldrich

Antimony(V) fluoride

for synthesis

Synonym(s):

Antimony(V) fluoride, Antimony pentafluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
F5Sb
CAS Number:
Molecular Weight:
216.75
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-021-8
NACRES:
NA.22
grade:
for synthesis
synthesis grade
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

bp

149-150 °C/1013 hPa

mp

7 °C

density

2.99 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/5F.Sb

InChI key

TWNMFECGADOEOJ-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service