Skip to Content
Merck
All Photos(1)

Key Documents

8.07064

Sigma-Aldrich

Pyrazine

for synthesis

Synonym(s):

Pyrazine, 1,4-Diazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2
CAS Number:
Molecular Weight:
80.09
MDL number:
UNSPSC Code:
12352005
EC Index Number:
206-027-6
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (GC)

form

crystals

bp

115-118 °C/1013 hPa

mp

49-52 °C

storage temp.

2-30°C

SMILES string

n1ccncc1

InChI

1S/C4H4N2/c1-2-6-4-3-5-1/h1-4H

InChI key

KYQCOXFCLRTKLS-UHFFFAOYSA-N

Application


  • X-ray transient absorption reveals the 1Au (npi*) state of pyrazine in electronic relaxation: This study reports on the photoinduced dynamics of pyrazine, providing insights into its electronic relaxation pathways (Scutelnic et al., 2021).

  • Theoretical evaluation of corrosion inhibition performance of some pyrazine derivatives: This paper evaluates the efficiency of pyrazine derivatives as corrosion inhibitors, providing a theoretical perspective on their adsorption properties (Obot and Gasem, 2014).

  • Pyrazine derivatives as green oil field corrosion inhibitors for steel: Investigates the effectiveness of pyrazine derivatives in preventing corrosion in oil field applications, emphasizing their environmentally friendly properties (Obot et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 51 °C
Melting range (upper value): ≤ 54 °C
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service