Skip to Content
Merck
All Photos(1)

Key Documents

8.06772

Sigma-Aldrich

4-Nitrobenzoyl chloride

for synthesis

Synonym(s):

4-Nitrobenzoyl chloride, 4-Nitrobenzoic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(NO2)C6H4COCl
CAS Number:
Molecular Weight:
185.56
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-517-4
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

solid

potency

5600 mg/kg LD50, oral (Rat)

bp

155 °C/20 hPa

mp

75 °C

storage temp.

2-30°C

SMILES string

ClC(=O)c1ccc(cc1)[N+](=O)[O-]

InChI

1S/C7H4ClNO3/c8-7(10)5-1-3-6(4-2-5)9(11)12/h1-4H

InChI key

SKDHHIUENRGTHK-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 97.0 %
Melting range (lower value): ≥ 70 °C
Melting range (upper value): ≤ 73 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service