Skip to Content
Merck
All Photos(1)

Documents

8.06749

Sigma-Aldrich

Nickel-aluminium alloy

(powder) for the production of Raney nickel for synthesis

Synonym(s):

Nickel-aluminium alloy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352402
NACRES:
NA.22

Quality Level

form

powder

mp

1460 °C

density

4.3 g/cm3

bulk density

2850 kg/m3

storage temp.

2-30°C

InChI

1S/Al.Ni

InChI key

NPXOKRUENSOPAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay of Ni: 47.0 - 53.0 %
Assay of Al: 47.0 - 53.0 %

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Sol. 2 - Skin Sens. 1 - STOT RE 1 - Water-react 3

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service