Skip to Content
Merck
All Photos(1)

Key Documents

8.06061

Sigma-Aldrich

2-Methoxyethyl acetate

for synthesis

Synonym(s):

2-Methoxyethyl acetate, Ethylene glycol monomethyl ether acetate, O-Methyl glycol acetate, 3-Oxabutyl acetate, Acetic acid 2-methoxyethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-772-9
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
144-145 °C/1013 hPa
vapor pressure:
4.7 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

4.7 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

380 °C (DIN 51794)

potency

2900 mg/kg LD50, oral (Rat)
5303 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-8.8 % (v/v)

dilution

(for synthesis)

bp

144-145 °C/1013 hPa

mp

-65 °C

transition temp

flash point 46 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOC(=O)C)C

InChI

1S/C5H10O3/c1-5(6)8-4-3-7-2/h3-4H2,1-2H3

InChI key

XLLIQLLCWZCATF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.004 - 1.007
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service