803071
Dimethyl sulfate
for synthesis
Synonym(s):
Dimethyl sulfate, Sulfuric acid dimethyl ester
About This Item
Recommended Products
vapor pressure
0.35 hPa ( 20 °C)
Assay
≥99% (GC)
form
liquid
autoignition temp.
450 °C
potency
106.4 mg/kg LD50, oral (Rat)
expl. lim.
3.6-23.2 % (v/v)
bp
188.5 °C/1013 hPa (decomposition)
mp
-31.8 °C
transition temp
flash point 83 °C
solubility
28 g/L (Hydrolysis)
density
1.33 g/cm3 at 20 °C
InChI
1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3
InChI key
VAYGXNSJCAHWJZ-UHFFFAOYSA-N
General description
Application
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
181.4 °F - closed cup
Flash Point(C)
83 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service