Skip to Content
Merck
All Photos(1)

Key Documents

8.03068

Sigma-Aldrich

N,N-Dimethylformamide dimethyl acetal

for synthesis

Synonym(s):

N,N-Dimethylformamide dimethyl acetal, N,N-Dimethyldimethoxymethylamine, DMF-DMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHN(CH3)2
CAS Number:
Molecular Weight:
119.16
MDL number:
UNSPSC Code:
12352116
EC Index Number:
225-063-3
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

liquid

autoignition temp.

155 °C

potency

>5000 mg/kg LD50, oral (Rat)

expl. lim.

1.3-17.7 % (v/v)

pH

7 ( in H2O)

mp

-85 °C

transition temp

flash point 6 °C

density

0.895 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+H](C(OC)OC)(C)C

InChI

1S/C5H13NO2/c1-6(2)5(7-3)8-4/h5H,1-4H3/p+1

InChI key

ZSXGLVDWWRXATF-UHFFFAOYSA-O

General description

N, N-Dimethylformamide dimethyl acetal (DMF-DMA) plays an important role in organic synthesis due to its higher reactivity.

  • As a methylating agent, DMF-DMA has been used in the synthesis of methyl esters from acids, methyl ethers, and thioethers from phenols and aromatic thiols.
  • DMF-DMA has been used as a formulating agent to synthesize enamines from active methylenes and active methyl groups, and amidines from amines and amides or thioamide.
  • It is also possible to produce heterocyclic compounds using DMF-DMA by cyclizing two functional groups.[1]

Application

N,N-Dimethylformamide dimethyl acetal is used:

  • In the synthesis of spirooxindole and pyrazole scaffolds via [3+2] cycloaddition reaction of electron-deficient olefins-based pyrazole motif.[2]
  • As a reagent in the synthesis of3-acetyl-4-arylquinoline-based enaminones under microwave irradiation.[3]

Analysis Note

Assay (HClO₄): ≥ 96.0 %
Density (d 20 °C/ 4 °C): 0.893 - 0.897
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethylformamide dimethyl acetal as a building block in heterocyclic synthesis
Abu-Shanab FA, et al.
Journal of Heterocyclic Chemistry, 46(5), 801-827 (2009)
Regio-and stereoselective synthesis of spiro-heterocycles bearing the pyrazole scaffold via [3+ 2] cycloaddition reaction
Journal of Molecular Structure, 1250, 131711-131711 (2022)
DMFDMA catalyzed synthesis of 2-((Dimethylamino) methylene)-3, 4-dihydro-9-arylacridin-1 (2H)-ones and their derivatives: in-vitro antifungal, antibacterial and antioxidant evaluations
Kumar L, et al.
open chemistry, 16(1), 1077-1088 (2018)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service