Skip to Content
Merck
All Photos(1)

Key Documents

8.02624

Sigma-Aldrich

2-Chloro-4-nitroaniline

for synthesis

Synonym(s):

2-Chloro-4-nitroaniline, 4-Amino-3-chloronitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(Cl)-4-(NO2)C6H3NH2
CAS Number:
Molecular Weight:
172.57
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-502-2

Assay

≥98.0% (HPLC)

autoignition temp.

522 °C

potency

6430 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 0.23 g/L in H2O)

bp

320 °C/1013 hPa (decomposes)

mp

104-107 °C

solubility

0.23 g/L

bulk density

850 kg/m3

storage temp.

2-30°C

InChI

1S/C6H5ClN2O2/c7-5-3-4(9(10)11)1-2-6(5)8/h1-3H,8H2

InChI key

LOCWBQIWHWIRGN-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 105 °C
Melting range (upper value): ≤ 108 °C
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service