Skip to Content
Merck

Skip To

8.02489

Citral

(mixture of cis- and trans-isomers) for synthesis

Synonym(s):

Citral, 3,7-Dimethyl-2,6-octadienal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
UNSPSC Code:
12352114
EC Index Number:
226-394-6
NACRES:
NA.22
MDL number:
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<1 hPa ( 50 °C)

Quality Segment

form

liquid

autoignition temp.

225 °C

potency

4950 mg/kg LD50, oral (Rat), 2250 mg/kg LD50, skin (Rabbit)

expl. lim.

4.3-9.9 % (v/v)

bp

227-229 °C/1013 hPa

mp

<-20 °C

transition temp

flash point 98 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C\C=C(\CCC=C(C)C)/C

InChI

1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3/b10-7+

InChI key

WTEVQBCEXWBHNA-JXMROGBWSA-N

Analysis Note

Assay isomers (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.887 - 0.889
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL82501C8300715861
Citral phyproof&#174; Reference Substance

PHL82501

Citral

Quality Level

200

Quality Level

-

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

-

storage temp.

2-30°C

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C

density

0.89 g/cm3 at 20 °C

density

0.888 g/mL at 25 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

mp

<-20 °C

mp

-

mp

-

mp

-

bp

227-229 °C/1013 hPa

bp

229 °C (lit.)

bp

229 °C (lit.)

bp

229 °C (lit.)


Still not finding the right product?

Explore all of our products under Citral


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

208.4 °F - DIN 51758

flash_point_c

98 °C - DIN 51758



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802489000504022536373713
802489025004022536373720
802489500004022536752075

Questions

Reviews

No rating value

Active Filters