Skip to Content
Merck
All Photos(1)

Key Documents

8.01504

Sigma-Aldrich

Silver acetate

for synthesis

Synonym(s):

Silver acetate, Acetic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3AgO2
CAS Number:
Molecular Weight:
166.91
MDL number:
UNSPSC Code:
12352103
EC Index Number:
209-254-9
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
crystals
solubility:
10.2 g/L

grade

for synthesis
synthesis grade

Quality Level

form

crystals

solubility

10.2 g/L

density

3.26 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Ag]OC(=O)C

InChI

1S/C2H4O2.Ag/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

CQLFBEKRDQMJLZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (from Ag): ≥ 98.0 %

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service