Skip to Content
Merck
All Photos(1)

Documents

8.00849

Sigma-Aldrich

Ethanolamine

for synthesis

Synonym(s):

Ethanolamine, 2-Aminoethanol, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7NO
CAS Number:
Molecular Weight:
61.08
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-483-3
NACRES:
NA.22

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

410 °C

potency

1515 mg/kg LD50, oral (Rat)
1025 mg/kg LD50, skin (Rabbit)

expl. lim.

3.4-27 % (v/v)

pH

12.1 (20 °C, 100 g/L in H2O)

kinematic viscosity

20 cSt(23 °C)

bp

171 °C/1013 hPa

mp

10.5 °C

transition temp

flash point 92.5 °C

density

1.02 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Application

  • Advancements in cyclotide structure and activity: Ethanolamine is explored within the context of reconstructing pseudo-ancestral cyclotides, which offers new insights into peptide stability and potential pharmaceutical applications (Jiang et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.015 - 1.016
Water (K. F.): ≤ 0.25 %
Sulfated ash: ≤ 0.1 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service