Skip to Content
Merck
All Photos(1)

Key Documents

8.00757

Sigma-Aldrich

Diethyl sebacate

for synthesis

Synonym(s):

Diethyl sebacate, Sebacic acid diethyl ester, Decanedioic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OOC(CH2)8COOC2H5
CAS Number:
Molecular Weight:
258.35
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-764-5

Quality Level

Assay

≥95.0% (GC)

potency

14470 mg/kg LD50, oral (Rat)

kinematic viscosity

6.2 cSt(20 °C)

bp

312 °C/1013 hPa

mp

-3 °C

transition temp

flash point 71 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C14H26O4/c1-3-17-13(15)11-9-7-5-6-8-10-12-14(16)18-4-2/h3-12H2,1-2H3

InChI key

ONKUXPIBXRRIDU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.963 - 0.966
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service