Skip to Content
Merck
All Photos(1)

Key Documents

8.00736

Sigma-Aldrich

4-Pyridinecarboxylic acid

for synthesis

Synonym(s):

4-Pyridinecarboxylic acid, iso-Nicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-228-2
NACRES:
NA.22
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

Assay

≥98% (HPLC)

form

powder

potency

5000 mg/kg LD50, oral (Rat)

pH

3-4 (20 °C, 6 g/L in H2O, saturated solution)

bp

260 °C/20 hPa

mp

315-319 °C

solubility

6 g/L

bulk density

500 kg/m3

storage temp.

no temp limit

SMILES string

[n+H]1ccc(cc1)C(=O)[O-]

InChI

1S/C6H5NO2/c8-6(9)5-1-3-7-4-2-5/h1-4H,(H,8,9)

InChI key

TWBYWOBDOCUKOW-UHFFFAOYSA-N

Related Categories

Application

  • N-sulfonic acid pyridinium-4-carboxylic acid chloride as a catalyst: This study describes the use of this compound as a catalyst for the condensation reaction of aldehyde with thiobarbituric acid. The catalytic performance is characterized in the context of improving reaction efficiency (Moosavi-Zare, Afshar-Hezarkhani, 2020).
  • Synthesis of Anti-inflammatory Substances: Focused on the synthesis of potential anti-inflammatory substances among triazole thioacetamides derived from 4-pyridinecarboxylic acid, this study expands on the pharmacological potentials of pyridine derivatives (Syrovaya, Chalenko, Demchenko, 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service