Skip to Content
Merck
All Photos(1)

Key Documents

8.00295

Sigma-Aldrich

Glutaric acid

for synthesis

Synonym(s):

Glutaric acid, Pentanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)3COOH
CAS Number:
Molecular Weight:
132.11
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-817-2
NACRES:
NA.22
form:
powder
Assay:
≥99.0% (acidimetric)
Pricing and availability is not currently available.

vapor pressure

0.022 hPa ( 18.5 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

powder

bp

302-304 °C/1013 hPa (slow decomposition)

mp

97.5-98 °C

transition temp

flash point 200 °C (DIN 51758)

density

1.43 g/cm3 at 15 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCC(=O)[O-].[H+].[H+]

InChI

1S/C5H8O4/c6-4(7)2-1-3-5(8)9/h1-3H2,(H,6,7)(H,8,9)

InChI key

JFCQEDHGNNZCLN-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 99,0 %(m)
Melting range (lower value): ≥ 95 °C
Melting range (upper value): ≤ 99 °C
Identity (IR): conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service