Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | $374.00 |
About This Item
manufacturer/tradename
BioControl Systems
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
for use with GDS Rotor-Gene
storage temp.
room temp
1 of 1
This Item | |||
|---|---|---|---|
| compatibility for use with GDS Rotor-Gene | compatibility for use with GDS Rotor-Gene | compatibility for use with GDS Rotor-Gene | compatibility for use with GDS Rotor-Gene® |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable |
| manufacturer/tradename BioControl Systems | manufacturer/tradename BioControl Systems | manufacturer/tradename BioControl Systems 73065 | manufacturer/tradename BioControl Systems |
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



