71335
pET-46 Ek/LIC Vector Kit - Novagen
pET-46 Ek/LIC Vector is a LIC-compatible derivative of pET-45b(+) containing a strong T7lac promoter and an amino-terminal His•Tag coding sequence immediately followed by an Ek/LIC cloning site.
Synonym(s):
pET System, pET-46 Ek/LIC Vector
About This Item
Recommended Products
Related Categories
General description
Components
- 1 µg- Ek/LIC Vector
- 25 units- T4 DNA Polymerase, LIC-qualified
- 50 µl- 25 mM EDTA
- 40 µl- 25 mM dATP
- 8 µl- Ek/LIC β-gal Control Insert
- 100 µl- 100 mM DTT
- 50 µl- 10X T4 DNA Polymerase Buffer
- 1.5 ml- Nuclease-free Water
- 5 × 2 ml- SOC Medium
- 10 µl- Test Plasmid
- 0.2 ml- BL21(DE3)pLysS Competent Cells
- 0.2 ml- BL21(DE3) Competent Cells
- 22 × 50 µl- NovaBlue GigaSingles Competent Cells
Warning
Other Notes
Legal Information
Disclaimer
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service