Skip to Content
Merck
All Photos(1)

Key Documents

688001

Sigma-Aldrich

Y-27632

≥95% (HPLC), liquid, Rho kinase inhibitor, Calbiochem®

Synonym(s):

InSolution Y-27632, Rho Kinase Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O · 2HCl · xH2O
Molecular Weight:
320.26 (anhydrous basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Y-27632, InSolution, ≥95%, reversible, inhibitor of Rho kinases

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A 5 mM (500 µg/296 µl) aqueous solution of Y-27632 (Cat. No. 688000). A highly potent, cell-permeable, selective inhibitor of Rho-associated protein kinase (Ki = 140 nM for p160ROCK (ROCK-I). Also inhibits ROCK-II with almost equal potency. Inhibition is achieved by competing with ATP for binding to the catalytic site. Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK. Does not affect the activity of p21-activated protein kinase (PAK) even at high concentrations (~100 mM). Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contractions (IC50 = 700 nM for phenylephrine-induced contractions in aortic strips).
A highly potent, cell-permeable, selective inhibitor of Rho-associated protein kinase (Ki = 140 nM for p160ROCK (ROCK-I). Also inhibits ROCK-II with almost equal potency. Inhibition is achieved by competing with ATP for binding to the catalytic site. Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK. Does not affect the activity of p21-activated protein kinase (PAK) even at high concentrations (~100 mM). Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contractions (IC50 = 700 nM for phenylephrine-induced contractions in aortic strips).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p160ROCK
Product competes with ATP.
Reversible: no
Target IC50: 700 nM against phenylephrine-induced contractions in aortic strips
Target Ki: 140 nM for p160ROCK (ROCK-I)

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

A 5 mM (500 µg/296 µl) solution of Y-27632 (Cat. No. 688000) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Chitaley, K., et al. 2001. Nat. Med.7, 119.
Davies, S.P., et al. 2000. Biochem. J. 351, 95.
Narumiya, S., et al. 2000. Methods Enzymol.325, 273.
Hirose, M., et al. 1999. J. Cell Biol.141, 1625.
Maekawa, M., et al. 1999. Science285, 895.
Uehata, M., et al. 1997. Nature389, 990.

Legal Information

Sold under license of PCT Application W098/06,433A1 and under license from Mitsubishi Pharma Tanabe Corporation.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvia J Gasparini et al.
The Journal of clinical investigation, 132(12) (2022-04-29)
Once human photoreceptors die, they do not regenerate, thus, photoreceptor transplantation has emerged as a potential treatment approach for blinding diseases. Improvements in transplant organization, donor cell maturation, and synaptic connectivity to the host will be critical in advancing this
Aaron C Brown et al.
Methods in molecular biology (Clifton, N.J.), 1926, 63-75 (2019-02-12)
Nephrons differentiate from the cap mesenchyme of the fetal kidney. Nephron progenitor cells that populate the cap mesenchyme efficiently balance self-renewal and epithelial differentiation to enable repeated rounds of nephron formation during development. Here we describe a method to isolate
Marina A Ayad et al.
Neurophotonics, 9(2), 025002-025002 (2022-06-03)
Significance: Interaction of neurons with their extracellular environment and the mechanical forces at focal adhesions and synaptic junctions play important roles in neuronal development. Aim: To advance studies of mechanotransduction, we demonstrate the use of the vinculin tension sensor (VinTS)
Radbod Darabi et al.
Methods in molecular biology (Clifton, N.J.), 1357, 423-439 (2014-11-19)
Cell-based therapies are considered as one of the most promising approaches for the treatment of degenerating pathologies including muscle disorders and dystrophies. Advances in the approach of reprogramming somatic cells into induced pluripotent stem (iPS) cells allow for the possibility
Rosita Bergström et al.
Methods in molecular biology (Clifton, N.J.), 767, 125-136 (2011-08-09)
Stem cell culture systems that rely on undefined animal-derived components introduce variability to the cultures and complicate their therapeutic use. The derivation of human embryonic stem cells and the development of methods to produce induced pluripotent stem cells combined with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service