Skip to Content
Merck
All Photos(1)

Key Documents

658390

Sigma-Aldrich

AG 9 - CAS 2826-26-8 - Calbiochem

Synonym(s):

[(4-Methoxybenzylidene)malononitrile, α-Cyano-(4-methoxy)cinnamonitrile], Tyrphostin A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥98% (TLC)

Quality Level

form

solid

color

yellow

solubility

ethanol: 10 mg/mL
DMSO: soluble
acetic acid: soluble

General description

An inactive compound that can be used as a negative control for inhibition of EGFR (IC50 >1250 μM for EGFR kinase). Has been shown to inhibit IL-2 stimulated Tyk-2 phosphorylation in ConA-activated T cells.

Biochem/physiol Actions

negative control for inhibition of EGFR

Target IC<SUB>50</SUB>:>1250 μM for EGFR kinase

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ruetten et al.
British journal of pharmacology, 122(1), 59-70 (1997-09-23)
1 Here we compared the effects of various inhibitors of the activity of protein tyrosine kinase on (i) the expression of the activity of the inducible isoform of nitric oxide (NO) synthase (iNOS) caused by endotoxin (lipopolysaccharide, LPS) in cultured
J J Bright et al.
Journal of immunology (Baltimore, Md. : 1950), 162(10), 6255-6262 (1999-05-07)
IL-12 is a macrophage-derived cytokine that induces proliferation, cytokine production, and cytotoxic activity of T and NK cells. Signaling through its receptor, IL-12 induces these cellular responses by tyrosine phosphorylation and activation of Janus kinase-2 (Jak-2), Tyk-2, Stat3, and Stat4.
A Gazit et al.
Journal of medicinal chemistry, 32(10), 2344-2352 (1989-10-01)
A novel class of low molecular weight protein tyrosine kinase inhibitors is described. These compounds constitute a systematic series of molecules with a progressive increase in affinity toward the substrate site of the EGF receptor kinase domain. These competitive inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service