Skip to Content
Merck

Skip To

650366-M

Tryptase, Human Lung

Sign In to View Organizational & Contract Pricing.

About This Item

EC Number:
NACRES:
NA.77
UNSPSC Code:
12352202
Specific activity:
≥50 units/mg protein


form

liquid

Quality Segment

specific activity

≥50 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

Native tryptase from human lung. Tryptase is a serine protease stored in mast cell granules. A tetrameric enzyme composed of four non-covalently bound subunits. Each subunit is reported to contain one active site. Reported to upregulate neoangiogenesis in carcinogenesis of the uterine cortex. Tryptase is also an important mediator of allergic reactions that activates G-protein-coupled receptors through proteolysis, thus amplifying histamine release.

Physical form

In 1 M NaCl, 50 mM NaOAc, 50 µM heparin, pH 5.0.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).
Prepared from tissue that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Other Notes

Benitez-Bribiesca, L., et al. 2001. J. Histochem. Cytochem.49, 1061.
He, S., et al. 1998. J. Pharmacol. Exp. Ther. 286, 289.
Molino, M., et al. 1997. J. Biol. Chem. 272, 4043.
Schwartz, L.B., and Bradford, T.R. 1986. J. Biol. Chem. 261, 7372.
One unit is defined as the amount of enzyme that gives a change of 1 absorbance per min at 410 nm using N-benzoyl-Phe-Val-Arg-pNA as a substrate in 30 min at 25°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters