Skip to Content
Merck
All Photos(2)

Documents

625625

Millipore

Trehalose, Dihydrate

Non-reducing disaccharide which yields 2 moles of D-glucose upon acid hydrolysis. Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro.

Synonym(s):

Trehalose, Dihydrate, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

Assay

≥97.5% (by Assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 100 mg/mL

cation traces

Fe: <5 ppm
heavy metals: <10 ppm

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

DPVHGFAJLZWDOC-PVXXTIHASA-N

General description

Non-reducing disaccharide that yields 2 moles of D-glucose upon acid hydrolysis. Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro. Oral administration of trehalose has been shown to effectively extend lifespan in a murine Huntington disease model. Also known to accumulate in osmotically-stressed cultured bacterial cells and protect against the adverse effects of dehydration. Reported to prevent myoglobin collapse and preserve its internal mobility.
Non-reducing disaccharide which yields 2 moles of D-glucose upon acid hydrolysis. Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro. Oral administration of trehalose has been shown to effectively extend lifespan in a murine Huntington disease model. Also known to accumulate in osmotically-stressed cultured bacterial cells and protect against the adverse effects of dehydration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Sarkar, S., et al. 2007. J. Biol. Chem. In press.
Tanaka, M., et al. 2004. Nat. Med.10, 148.
Sastry, G.M. and Agmon, N. 1997. Biochemistry 36, 7097.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service