Skip to Content
Merck
All Photos(1)

Documents

615700

Sigma-Aldrich

Total Antioxidant Status Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.84

usage

sufficient for 50 tests

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

assay range

standard curve range: 0-2.5 mM
(plasma)

input

sample type serum
sample type plasma
sample type tissue extract(s)
sample type beverage(s)
sample type plant extract(s)

detection method

colorimetric

shipped in

ambient

storage temp.

2-8°C

General description

A colorimetric assay kit for the measurement of the total antioxidant status. Each kit is suitable for performing a minimum of 50 assays.
Note: Spectrophotometer equipped with a temperature-controlled cuvette holder is strongly recommended for use with this kit.

Components

Substrate, Chromogen, Standard, Buffer, and a user protocol.

Warning

Toxicity: Standard Handling (A)

Principle

The Calbiochem Total Antioxidant Status Assay Kit is designed to assay antioxidant levels in serum or plasma samples. Additionally, the assay may be used to measure the antioxidant potential of (suitably solubilized) food and drug samples.

Preparation Note

• Chromogen: Add 10 ml Buffer to each vial of Chromogen needed for the assay (10 tests per vial). After reconstitution, the Chromogen is stable for 2 days at 4°C or 8 h at room temperature.

• Substrate: Dilute each vial of Substrate needed for the assay by adding 7.5 ml Buffer. After dilution, the substrate is stable for 24 h at 4°C. NOTE: Some automated methods do not require substrate dilution.

• Standard: Add 1 ml distilled water to each vial of Standard needed for the assay. After reconstitution, the Standard is stable for 2 days at 4°C or 1 month at -20°C.

Storage and Stability

Upon arrival store the entire kit contents at 4°C. Note: The concentration of the standard is lot-specific. Please refer to the vial label for lot-specific concentration.

Analysis Note

Calculation of Total Antioxidant StatusThe antioxidant concentration of the Standard is used to calculate the antioxidant levels in the samples.

1. Determine the ΔA for the samples, standard, and blank. ΔA = A - Ao
2. Calculate the concentration of antioxidants in the sample using the formula below:
<div class="Bio_doc_image">Figure 2: Antioxidant Concentration Formula
</div>
NOTE: If the antioxidant concentration is greater than 2.5 mM dilute the sample with 0.9% NaCl and re-assay. Typical antioxidant levels in normal human plasma are 1.30-1.77 mM.

Other Notes

Miller, N.J., et al. 1993. Clin. Science 84, 407.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rori Morrow et al.
Journal of medicinal food, 12(2), 298-303 (2009-05-23)
Oxidative stress and inflammation have been linked to bone loss. We evaluated the effects of feeding orange pulp (OP), a source of vitamin C and flavonoids, on bone quality in a rat model of male osteoporosis. One-year-old retired breeder rats
Jean Michell Santoyo et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
Oxidative and inflammatory stress, angiogenic imbalance, and endothelial dysfunction are pathophysiological mechanisms occurring in pre-eclampsia (PE) that may persist over time and predispose women to a higher risk of cardiovascular disease (CVD) in the future. However, there is little evidence
M Vázquez-Añón et al.
Journal of dairy science, 91(8), 3165-3172 (2008-07-25)
The objective of the study was to evaluate the effect of feeding the dietary antioxidant Agrado Plus (AOX; Novus International, St. Louis, MO) in diets that contained 2% fresh fat (FF) or oxidized fat (OF) on milk production and composition
Hui Eng Leh et al.
Annals of medicine, 53(1), 1059-1065 (2021-06-29)
The use of lycopene as a complementary medicine for Type II diabetes mellitus (T2DM) is limited and controversial. This study evaluated the effect of lycopene intake on the changes of glycaemic status and antioxidant capacity among the T2DM patients. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service