Skip to Content
Merck
All Photos(1)

Key Documents

6.08685

Supelco

Xylene

Synonym(s):

Xylenes, Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12131501
bp:
137-140 °C (lit.)
vapor pressure:
18 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

vapor pressure

18 mmHg ( 37.7 °C)

expl. lim.

7 %

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service