Skip to Content
Merck
All Photos(1)

Documents

5874316024C

Milli-Q

EfferSan Effervescent Tablets

Protects the reverse osmosis (RO) membrane from contaminants

Synonym(s):

EfferSan Effervescent Tablets (CA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104210
eCl@ss:
33050190
NACRES:
JA.13

Quality Level

form

tablet

feature

Formerly known as ROProtect CR

composition

(mixture of troclosene sodium dihydrate, sodium carbonate and adipic acid)

packaging

pkg of 24 tablets

availability

available only in USA and Canada

storage condition

dry at room temperature (Tightly closed. Do not store near acids)

H

10.2 cm (4 in.)

compatibility

for use with AFS®
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Essential
for use with Elix® Reference
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® HR
for use with Milli-Q® HX
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
for use with RiOs Essential

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

EfferSan Effervescent Tablets (CA) are chlorine tablets used in filtration devices such as water purification systems and cartridges. This product optimally protects reverse osmosis (RO) membranes from fouling contaminants.

Application

EfferSan Tablets are used for research and development purposes such as biochemical research/analysis or for application as an analytical reagent.

Features and Benefits

Prevents bacterial proliferation and biofilm formation to preserve RO membrane performance (ionic rejection, water recovery).

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Desen. Expl. 4 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.

Related Content

Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service