Skip to Content
Merck
All Photos(1)

Key Documents

580567

Millipore

TCEP, Hydrochloride, Reagent Grade

Synonym(s):

TCEP, Hydrochloride, Reagent Grade, tris(2-Carboxyethyl)phosphine, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15O6P · xHCl
CAS Number:
Molecular Weight:
250.19 (free base basis)
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.25
form:
solid
Assay:
≥98% (GC)
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

≥98% (GC)

form

solid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

P(CCC(=O)[O-])(CCC(=O)[O-])CCC(=O)[O-]

InChI

1S/C9H15O6P/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15/h1-6H2,(H,10,11)(H,12,13)(H,14,15)/p-3

InChI key

PZBFGYYEXUXCOF-UHFFFAOYSA-K

General description

A water-soluble and odorless disulfide-reducing agent that is more stable and effective than DTT. TCEP reduces even the most stable water-soluble alkyl disulfides at pH 4.5 within 5 minutes. TCEP is also capable of functioning as a reducing agent even in the pH range of 1.5-8.5. It is more resistant to air-oxidation as compared to DTT.

Application


  • A Study on Thiol-Michael Addition to Semi-Synthetic Elastin-Hyaluronan Material for Electrospun Scaffolds.: Investigates the application of TCEP hydrochloride in the thiol-Michael addition reactions to develop semi-synthetic elastin-hyaluronan materials, which are pivotal for creating electrospun scaffolds used in biomedical engineering (Laezza et al., 2024).

  • Target Recognition-Triggered Peroxidase-Mimicking Activity Depression in Homochiral Nanochannels for Identifying Cystine Enantiomers.: Highlights the use of TCEP hydrochloride in the development of a novel method for identifying cystine enantiomers through target recognition-triggered peroxidase-mimicking activity depression in nanochannels (Xu et al., 2023).

Packaging

Packaged under inert gas

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). TCEP retains stability when stored at an acidic or alkaline pH; avoid storage at pH 7.0-8.0. Also avoid exposure to high concentrations of phosphate ions (>150 mM). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Getz, E.B., et al. 1999. Anal. Biochem. 273, 73.
Han, J.C. and Han, G.Y. 1994. Anal. Biochem. 220, 5.
Burns, J.A., et al. 1991. J. Org. Chem.56, 2648.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinzhao Song et al.
Analytical chemistry, 93(38), 13063-13071 (2021-09-21)
Short of a vaccine, frequent and rapid testing, preferably at home, is the most effective strategy to contain the COVID-19 pandemic. Herein, we report on single-stage and two-stage molecular diagnostic tests that can be carried out with simple or no

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service