Skip to Content
Merck

574591

Superoxide Dismutase, Human, Recombinant, E. coli

Synonym(s):

5-LO, Cu/Zn SOD, Human, Recombinant, E. coli

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Specific activity:
≥3000 U/mg
Assay:
≥90% (SDS-PAGE)
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

assay

≥90% (SDS-PAGE)

form

solid

specific activity

≥3000 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

soluble (Reconstitute in sterile MQ water to a final concentration of >=100 µg/ml. Further dilute in aqueous buffers just prior to use.)

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human superoxide dismutase expressed in E. coli as a homodimer consisting of 2 identical, 154-amino acid chains. Purified by proprietary chromatographic methods. Superoxide dismutase is an essential component of biological defense against the toxic effects of oxygen. It catalyzes the dismutation of superoxide radicals to molecular oxygen and H2O2 and plays a role in cellular aging due to its reduced activity in aging cells. Mutant forms of Cu/Zn SOD are associated with neurodegenerative disease.

Physical form

Lyophilized from a 0.2μm filtered concentrated (1mg/ml) solution in PBS, pH 7.4.

Preparation Note

Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 2 weeks at 4°C or for up to 4 months at -20°C.
Reconstitute in sterile MQ water to a final concentration of ≥100 µg/ml. Further dilute in aqueous buffers just prior to use.

Other Notes

Baumber, J. and Ball, B.A. 2005. Am. J. Vet. Res.66, 1415.
Dimitrova, A.A., et al. 2005. Folia Med. (Plovdiv)47, 42.
Jebara, S., et al. 2005. J. Plant Physiol.162, 929.
Mila-Kierzenkowska, C., et al. 2005. Acta Biol. Hung.56, 399.
Miszalski, Z., et al. 2005. J. Plant Physiol.162, 895.
Paramonov, B.A., et al. 2005. Bull. Exp. Biol. Med.139, 56.
≥3000 units/mg.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library