Skip to Content
Merck
All Photos(1)

Documents

565834

Sigma-Aldrich

sFRP-3, His•Tag®, Mouse, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse secreted frizzled related protein 3 (sFRP-3) consisting of amino acids Ala 33-Ser323, fused at the N-terminus to a His•Tag sequence and expressed in NSO-derived mouse myeloma cells. sFRP-3 is a soluble antagonist of Wnt signaling. It prohibits interactions with Frizzled receptors by binding directly to Wnt proteins.

Biochem/physiol Actions

Proliferation Assay:

ED₅₀ = 1.6-6 µg/ml as measured by inhibition of proliferation of MC3T3-E1 mouse osteoblastic cells.



Functional ELISA:

50% optimal binding response = 150-250 ng/ml as measured by binding ro rmWnt-3a (500 ng/ml).

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered PBS, BSA.

Reconstitution

Following reconstituion refrigerate (4°C) for short term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute with sterile PBS contanining ≥1% human or bovine serum albumin. Please refer to the vial label for lot-specific reconstitution volume.

Other Notes

Hoang, et al. 1996. J. Biol. Chem.271, 26131.Leyns, et al. 1997 Cell88, 747.Wang, et al. 1997 Cell88, 757.Mayr, et al. 1997 Mech. Dev.63, 109.Rattner, et al. 1997. PNAS94, 2859.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service