Skip to Content
Merck
All Photos(1)

Documents

557368

Sigma-Aldrich

Rotenone

≥98% (HPLC), solid, NADH-CoQ reductase inhibitor, Calbiochem

Synonym(s):

Rotenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Rotenone, A mitochondrial toxin and a potent, reversible, and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

ethanol: 5 mg/mL
DMSO: 50 mg/mL
chloroform: 50 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C23H22O6/c1-11(2)16-8-14-15(28-16)6-5-12-22(24)21-13-7-18(25-3)19(26-4)9-17(13)27-10-20(21)29-23(12)14/h5-7,9,16,20-21H,1,8,10H2,2-4H3/t16-,20-,21+/m1/s1

InChI key

JUVIOZPCNVVQFO-HBGVWJBISA-N

General description

A mitochondrial toxin and a potent, reversible and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain. Also inhibits cellular proliferation in mouse liver.
A mitochondrial toxin and a potent, reversible, and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain. Also inhibits cellular proliferation in mouse liver.

Biochem/physiol Actions

Cell permeable: no
Primary Target
NADH-CoQ reductase
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot, flush with nitrogen, and store at -70°C. Stock solutions are stable for up to 1 month at -70°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kopustinskiene, D.M., et al. 2001. Eur. J. Pharmacol.428, 311.
Higgins, D.S., Jr. and Greenamyre, J.T. 1996. J. Neurosci. 16, 3807.
Cunningham, M.L., et al. 1995. Cancer Lett. 95, 93.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andaleb Kholmukhamedov et al.
The Journal of biological chemistry, 298(9), 102336-102336 (2022-08-06)
Mitochondrial chelatable iron contributes to the severity of several injury processes, including ischemia/reperfusion, oxidative stress, and drug toxicity. However, methods to measure this species in living cells are lacking. To measure mitochondrial chelatable iron in living cells, here we synthesized
Tongling Liufu et al.
Frontiers in physiology, 14, 1164287-1164287 (2023-08-31)
Introduction: Mitochondrial disease is a spectrum of debilitating disorders caused by mutations in the mitochondrial DNA (mtDNA) or nuclear DNA that compromises the respiratory chain. Mitochondrial 3243A>G (m.3243 A>G) is the most common mutation showing great heterogeneity in phenotype. Previous
Courtney J Banks et al.
Molecular and cellular biology, 37(20) (2017-07-26)
In this study, we employed proteomics to identify mechanisms of posttranslational regulation on cell survival signaling proteins. We focused on Cu-Zn superoxide dismutase (SOD1), which protects cells from oxidative stress. We found that acylation of K122 on SOD1, while not
John J Wilson et al.
iScience, 26(9), 107487-107487 (2023-08-28)
Aberrant metabolic demand is observed in immune/inflammatory disorders, yet the role in pathogenesis remains unclear. Here, we discover that in lupus, activated B cells, including germinal center B (GCB) cells, have remarkably high glycolytic requirement for survival over T cell populations
Travis R Madaris et al.
Cell reports, 42(3), 112155-112155 (2023-03-02)
The most abundant cellular divalent cations, Mg2+ (mM) and Ca2+ (nM-μM), antagonistically regulate divergent metabolic pathways with several orders of magnitude affinity preference, but the physiological significance of this competition remains elusive. In mice consuming a Western diet, genetic ablation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service