Skip to Content
Merck
All Photos(1)

Key Documents

554994

Sigma-Aldrich

RHC-80267

Selective inhibitor of DAG lipase activity in canine platelets (IC₅₀ = 4 nM) and in a variety of mammalian cells.

Synonym(s):

RHC-80267, 1,6- bis(Cyclohexyloximinocarbonylamino)hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34N4O4
CAS Number:
Molecular Weight:
394.51
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C20H34N4O4/c25-19(27-23-17-11-5-3-6-12-17)21-15-9-1-2-10-16-22-20(26)28-24-18-13-7-4-8-14-18/h1-16H2,(H,21,25)(H,22,26)

InChI key

RXSVYGIGWRDVQC-UHFFFAOYSA-N

General description

Selective inhibitor of DAG lipase activity in canine platelets (IC50 = 4 nM) and in a variety of mammalian cells. Also inhibits glucose- and carbachol-induced insulin release from intact islets. An inhibitor of Angiotensin II (Cat. No. 05-23-0101) and ATP-induced synthesis of 6-keto-prostaglandin F1α.

Biochem/physiol Actions

Cell permeable: no
Primary Target
DAG lipase
Product does not compete with ATP.
Reversible: no
Target IC50: 4 nM against DAG lipase activity in canine platelets

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shinoda, J., et al. 1997. Arterioscler. Thromb. Vasc. Biol. 17, 295.
Shinoda, J., et al. 1997. Eur. J. Endocrinol. 136, 207.
Konrad, R.J., et al. 1994. Biochemistry 33, 13284.
Mason-Garcia, M., et al. 1992. Am. J. Physiol.262, C1197.
Balsinde, J., et al. 1991. J. Biol. Chem.266, 15638.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service