Skip to Content
Merck
All Photos(1)

Documents

553506-M

Sigma-Aldrich

RAP, Human, Recombinant, E. coli

Synonym(s):

Receptor Associated Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

Recombinant, human RAP fused to GST, expressed in E. coli, and subsequently cleaved with thrombin to remove GST. Antagonizes the ligand binding of certain members of the low-density lipoprotein receptor family such as low density lipoprotein receptor-related protein (LRP) and very low-density lipoprotein (VLDL) receptor. Promotes the proper folding and subsequent exocytic trafficking of the wild-type low density lipoprotein receptor (LDLR) and several of its class 2 mutants. Expressed as a GST-fusion protein that is subsequently cleaved with thrombin. Typical concentrations to block LRP or VLDL receptor are 200-300 nM. For antagonism of LDLR, 1 µM is recommended.
Recombinant, human RAP fused to GST, expressed in E. coli, and subsequently cleaved with thrombin to remove GST. Receptor Associated Protein (RAP) antagonizes the ligand binding of certain members of the low-density lipoprotein receptor family such as low-density lipoprotein receptor-related protein (LRP) and very low density lipoprotein (VLDL) receptor. Promotes the proper folding and subsequent exocytic trafficking of the wild-type low-density lipoprotein receptor (LDLR) and several of its class 2 mutants. Typical concentrations used to block LRP or VLDL receptor are 200-300 nM; for antagonism of LDLR, 1 µM is recommended.

Warning

Toxicity: Standard Handling (A)

Physical form

150 mM NaCl, 100 mM Tris-HCl, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Li, Y., et al. 2002. Biochemistry41, 4921.
Salicioni, A.M., et al. 2002. J. Biol. Chem.277, 16160.
Anderson, O.M., et al. 2001. Biochemistry40, 15408.
Bu, G. 2001. Int. Rev. Cytol.209, 79.
Williams, S.E., et al. 1992. J. Biol. Chem.267, 9035

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service