Skip to Content
Merck
All Photos(1)

Key Documents

5.43897

Supelco

N,N-Dimethylformamide

for liquid chromatography LiChrosolv®

Synonym(s):

DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$208.00
2.5 L
$454.00

$208.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
1 L
$208.00
2.5 L
$454.00

About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-679-5
NACRES:
NA.03
technique(s):
HPLC: suitable
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)

$208.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

description

isocratic

product line

LiChrosolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

technique(s)

HPLC: suitable

impurities

≤0.03% Water

transmittance

275 nm, ≥65%
300 nm, ≥90%
350 nm, ≥99.5%
400 nm, ≥99.5%

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

solubility

1000 g/L

density

0.944 g/mL (lit.)

storage temp.

2-30°C

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent for many hydrophobic organic compounds.

Analysis Note

Purity (GC): ≥ 99.9 %
Water: ≤ 0.03 %
Transmission (at 275 nm): ≥ 65 %
Transmission (at 300 nm): ≥ 90 %
Transmission (at 350 nm): ≥ 99.5 %
Transmission (at 400 nm): ≥ 99.5 %

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service