Skip to Content
Merck
All Photos(1)

Documents

539160

Sigma-Aldrich

Proteasome Inhibitor I

The Proteasome Inhibitor I controls the biological activity of Proteasome. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Proteasome Inhibitor I, PSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H50N4O8
Molecular Weight:
618.76
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 30 mg/mL
ethanol: 30 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible inhibitor of the chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells. Causes the accumulation of ubiquitinated proteins in neuronal cells. Prevents the activation of NF-κB in response to TNF-α and Okadaic Acid (Cat. No. 495604) through inhibition of IκB-α degradation. A 50 mM (5 mg/162 µl) solution of Proteasome Inhibitor I (Cat. No. 539161) in DMSO is also available.
Cell-permeable inhibitor of the chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells. Causes the accumulation of ubiquitinylated proteins in neuronal cells. Prevents NF-κB activation in response to TNFα and okadaic acid through inhibition of IκB-α degradation, thereby interfering with induction of iNOS in macrophages.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells
Product does not compete with ATP.
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Ile-Glu(OtBu)-Ala-Leu-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 3 months at -20°C.

Other Notes

Griscavage, J.M., et al. 1996. Proc. Natl. Acad. Sci. USA93, 3308.
Mori, S., et al. 1995. J. Biol. Chem.270, 29447.
Figueiredo-Pereira, M.E., et al. 1994. J. Neurochem. 63, 1578.
Traenckner, E.B., et al. 1994. EMBO J. 13, 5433.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service